missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ 93-143 Disposable Nitrile Gloves with Fully Textured Grip

Product Code. 16895430
Click to view available options
6
This item is not returnable. View return policy
This item is not returnable. View return policy

Versatile gloves offer reliable, medium-duty protection with a textured palm to enhance grip capabilities in wet or oily applications.

  • Ideal for a wide variety of industrial applications
  • Textured palm enhances grip in wet, dry, and oily environments
  • Silicone-free for better product protection
  • Tested for use with chemotherapy drugs
TRUSTED_SUSTAINABILITY

Specifications

Chemotherapeutic Rated Yes
AQL 1.5
Color Blue
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 6
Thickness (English) 4.7 mil
Thickness (Metric) 0.12 mm
Glove Cuff Style Beaded
Length (English) 9.5 in.
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'