missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell MICROCHEM™ 6000
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
O60A812E
O60A813E
O60A818E
O60A819E
O60T801B
O60T802E
O60T803E
O60T808E
O60T809E
Description
- Excellent chemical protection, >480 mins for 15 chemicals specified by EN 943-2:2002
- Innovative fabric technology, high performance barrier laminate with scrim reinforcement
- Lightweight and flexible
- Highly visible bright orange color for improved worker safety
- Ultrasonically welded and hot air taped seams
- DYNAT/YKK Gas-Tight zipper provides protection and performance in the most hostile of chemical environments
- The zip has an outer zip flap made of the same material as the suit
- Semi-rigid 3 layer GAG visor provides excellent optical clarity
Specifications
Specifications
Color | Orange |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning |
Material | Multi-Layer Non-Woven Barrier Laminate Fabric |
Size | 2X-Large |
Seam Type | Welded and Taped |
Style | O60A813E |
Weight (Metric) | 230 gsm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction