Learn More
Fisherbrand™ Micro Insert for well plates, glass
For complex UHPLC, LC and GC applications.
3370.00 NOK - 4040.00 NOK
Specifications
Color Clear
Material Glass
View More Specs
Products 4
Product Code Brand Volume (Metric) Style Price Quantity & Availability  
Product Code Brand Volume (Metric) Style Price Quantity & Availability  
10091762
missing translation for 'viewDocuments'
Fisherbrand™
08 09 1081
1000 ÎĽL Constricted
4040.00 NOK
Pack of 960
Only null left
In Stock
Add to basket
10275772
missing translation for 'viewDocuments'
Fisherbrand™
08 09 0909
1200 ÎĽL Round
3470.00 NOK
Pack of 960
Only null left
In Stock
Add to basket
10377552
missing translation for 'viewDocuments'
Fisherbrand™
08 09 0903
1000 ÎĽL Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10295072
missing translation for 'viewDocuments'
Fisherbrand™
08 09 0888
400 ÎĽL Tapered
3370.00 NOK
Pack of 960
Only null left
In Stock
Add to basket
Description
Description
  • Clear
  • Glass
  • Plates meet ANSI and SBS standards
  • Different sealing options
  • Chromatography tested
  • No leeching due to high-end material
Specifications
Specifications
Clear
Glass
Product Title
Fisherbrand™ Micro Insert for well plates, glass