missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15772-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5′ UTR may exist, but their biological validity has not been determined.Specifications
MGAT2 | |
Polyclonal | |
Unconjugated | |
MGAT2 | |
CDGS2, GlcNAc T II, GLCNACTII, GNT II, GNT2, MGAT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217664, 4247 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q10469, Q921V5 | |
MGAT2 | |
MGAT2 Fusion Protein Ag8423 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction