Learn More
METTLER TOLEDO™ SevenExcellence™ S470 pH/Conductivity Benchtop Meter
Tienda Mettler Toledo Productos
Click to view available options
Model:
Meter, Standard Kit
Meter, USP/EP Compliance Kit
S470 pH/Conductivity Meter
Tamaño de la unidad:
Each
Descripción
- pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET meter than can be expanded with additional measurement modules to measure dissolved oxygen, ISE, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
Especificaciones
Especificaciones
| Outputs | RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA |
| Accuracy (Conductivity) | ±0.5% |
| Accuracy (mV) | ±0.1mV |
| Accuracy (pH) | ±0.002 |
| Calibration Points (Conductivity) | 13 Pre defined and 20 user defined standard |
| Calibration Points (ISE) | Max. 5 points, 8 pre defined and 20 user defined buffer group |
| Calibration Points (pH) | Max. 5 points, 8 pre defined and 20 user defined buffer group |
| Data Points | 20,000 |
| Display Type | TFT |
| Range (Conductivity) | 0.001 μS/cm to 2000 mS/cm |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido