Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Click to view available options
Dimensions (L x W):
35.5 x 182.9 in. (14 x 72 cm)
36 x 60 in. (91 x 152.4 cm)
45.7 x 121.9 in. (18 x 48 cm)
45.7 x 183 in. (18 x 72 cm)
45.7 x 61 in. (18 x 24 cm)
53 x 106.6 in. (21 x 42 cm)
53 x 21.9 in. (21 x 48 cm)
53 x 63 in. (28 x 28 cm)
53 x 76 in. (30 x 30 cm)
61 x 106.6 in. (24 x 42 cm)
61 x 121.9 in. (24 x 48 cm)
61 x 137 in. (24 x 54 cm)
61 x 152.4 in. (24 x 60 cm)
76 x 121.9 in. (30 x 48 cm)
76 x 182.9 in. (30 x 72 cm)
76 x 91.4 in. (30 x 36 cm)
91 x 182.9 in. (36 x 72 cm)
91 x 91 in. (36 x 36 cm)
Depth (Metric):
355 mm
457 mm
530 mm
610 mm
760 mm
910 mm
Unit Size:
Each
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Type 304 polished stainless steel
- Used for the most aggressive applications and environments
- Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
Product Type | Wire Shelf |
Material | Stainless Steel |
Depth (English) | 36 in. |
Depth (Metric) | 910 mm |
Dimensions (L x W) | 91 x 91 in. (36 x 36 cm) |
For Use With (Equipment) | Metro Super Erecta Shelving System |
Includes | Plastic sleeves and wedges |
Product Content Correction
Product Title
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish >