Learn More
Metro™ Super Erecta™, Super Erecta Pro™ and Super Adjustable™ 2 Top-Track Stationary End Unit Kits
Connects stationary end units to overhead track sets.
9505.00 NOK - 16445.00 NOK
Specifications
For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system |
---|---|
Includes | 86 in. Posts and connecting hardware |
Product Type | Stationary End Unit Kit |
Product Code | Brand | Material | Width (English) Shelf | Finish | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Width (English) Shelf | Finish | Price | Quantity & Availability | ||||
15927291
|
Metro™
TTE18S |
Stainless Steel | 18 in. | None |
16055.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
15957291
|
Metro™
TTE21S |
Stainless Steel | 21 in. | None |
16260.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
15987291
|
Metro™
TTE24S |
Stainless Steel | 24 in. | None |
16445.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
15907291
|
Metro™
TTE18C |
Steel | 18 in. | Chrome |
9505.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
15937291
|
Metro™
TTE21C |
Steel | 21 in. | Chrome |
9695.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
15967291
|
Metro™
TTE24C |
Steel | 24 in. | Chrome |
9880.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||
15917291
|
Metro™
TTE18K3 |
Steel | 18 in. | Metroseal 3 with Microban™ |
9880.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
ÂżYa tiene una cuenta? Inicie sesiĂłn para ver su precio y disponibilidad. ÂżEs nuevo aquĂ? ¡RegĂstrese hoy mismo! |
|||
15947291
|
Metro™
TTE21K3 |
Steel | 21 in. | Metroseal 3 with Microban™ |
10015.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
ÂżYa tiene una cuenta? Inicie sesiĂłn para ver su precio y disponibilidad. ÂżEs nuevo aquĂ? ¡RegĂstrese hoy mismo! |
|||
15977291
|
Metro™
TTE24K3 |
Steel | 24 in. | Metroseal Green Epoxy |
10220.00 NOK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
ÂżYa tiene una cuenta? Inicie sesiĂłn para ver su precio y disponibilidad. ÂżEs nuevo aquĂ? ¡RegĂstrese hoy mismo! |
|||
DescripciĂłn
- Kit includes 86 in. (218.5cm) posts and hardware necessary for connecting stationary end units to track
- Super Erecta Shelves open-wire design minimizes dust accumulation, allows a free circulation of air and greater visibility of stored items.
MetroSeal 3 Finish is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
InformaciĂłn de Ăłrdenes
One kit is requird per single system. Shelves sold separately.
Especificaciones
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system | |
86 in. Posts and connecting hardware | |
Stationary End Unit Kit |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.