Learn More
Emma Safety Footwear Metric Safety Shoes
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
1225.00 NOK - 1460.00 NOK
Specifications
Certifications/Compliance | EN ISO 20345:2011, EN 12568:2010 |
---|---|
Closure Type | Elastic |
Color | White |
Design | Metric |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15928541
|
Emma Safety Footwear
MM50654035 |
35 |
1225.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15938541
|
Emma Safety Footwear
MM50654036 |
36 |
1225.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15948541
|
Emma Safety Footwear
MM50654037 |
37 |
1225.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15958541
|
Emma Safety Footwear
MM50654038 |
38 |
1225.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15968541
|
Emma Safety Footwear
MM50654039 |
39 |
1460.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15978541
|
Emma Safety Footwear
MM50654040 |
40 |
1355.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15988541
|
Emma Safety Footwear
MM50654041 |
41 |
1280.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15998541
|
Emma Safety Footwear
MM50654042 |
42 |
1420.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15908551
|
Emma Safety Footwear
MM50654043 |
43 |
1315.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15918551
|
Emma Safety Footwear
MM50654044 |
44 |
1420.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15928551
|
Emma Safety Footwear
MM50654045 |
45 |
1225.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15938551
|
Emma Safety Footwear
MM50654046 |
46 |
1225.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15948551
|
Emma Safety Footwear
MM50654047 |
47 |
1365.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15958551
|
Emma Safety Footwear
MM50654048 |
48 |
1420.00 NOK
1 pair |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Easy Twist Outsole
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
White | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Elastic | |
Metric | |
Microfiber |