missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-chloropyridazine-4-carboxylate, 97%, Thermo Scientific Chemicals

2092.79 NOK - 5643.49 NOK

Chemical Identifiers

CAS 1093860-48-0
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD16660968
InChI Key QYQHGLKSLGUINZ-UHFFFAOYSA-N
Synonym 6-chloro-pyridazine-4-carboxylic acid methyl ester, methyl6-chloropyridazine-4-carboxylate, 4-pyridazinecarboxylicacid, 6-chloro-, methyl ester, 6-chloro-4-pyridazinecarboxylic acid, methyl ester, 4-pyridazinecarboxylic acid, 6-chloro-, methyl esterShow MoreShow Less
PubChem CID 45790080
IUPAC Name methyl 6-chloropyridazine-4-carboxylate
SMILES COC(=O)C1=CC(Cl)=NN=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10098289
SDS View Documents
Thermo Scientific Acros
432980010
1 g Glass bottle
2092.79 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
10331230
SDS View Documents
Thermo Scientific Acros
432980050
5 g Glass bottle
5643.49 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 6-chloropyridazine-4-carboxylate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.