Learn More
Methyl trans-2-pentenoate, 95%, Thermo Scientific Chemicals
1068.74 NOK - 3374.34 NOK
Chemical Identifiers
CAS | 15790-88-2 |
---|---|
Molecular Formula | C6H10O2 |
Molecular Weight (g/mol) | 114.14 |
MDL Number | MFCD00137611 |
InChI Key | MBAHGFJTIVZLFB-SNAWJCMRSA-N |
Synonym | methyl 2-pentenoate, methyl trans-2-pentenoate, methyl 2e-pent-2-enoate, 2-pentenoic acid, methyl ester, 2-pentenoic acid, methyl ester, e, methyl 3-ethylacrylate, methyl 2-transpentenoate, methyl 2-trans-pentenoate, methyl e-2-pentenoate, trans-methyl pent-2-enoate |
PubChem CID | 5364718 |
SMILES | CC\C=C\C(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10662192
|
Thermo Scientific
329720050 |
5 g | Plastic bottle |
1068.74 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10111091
|
Thermo Scientific Acros
329720250 |
25 g | Plastic bottle |
3374.34 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15790-88-2 | |
114.14 | |
MBAHGFJTIVZLFB-SNAWJCMRSA-N | |
5364718 |
C6H10O2 | |
MFCD00137611 | |
methyl 2-pentenoate, methyl trans-2-pentenoate, methyl 2e-pent-2-enoate, 2-pentenoic acid, methyl ester, 2-pentenoic acid, methyl ester, e, methyl 3-ethylacrylate, methyl 2-transpentenoate, methyl 2-trans-pentenoate, methyl e-2-pentenoate, trans-methyl pent-2-enoate | |
CC\C=C\C(=O)OC |
Specifications
15790-88-2 | |
20°C | |
94% min. (GC) | |
1.4300 to 1.4320 | |
methyl 2-pentenoate, methyl trans-2-pentenoate, methyl 2e-pent-2-enoate, 2-pentenoic acid, methyl ester, 2-pentenoic acid, methyl ester, e, methyl 3-ethylacrylate, methyl 2-transpentenoate, methyl 2-trans-pentenoate, methyl e-2-pentenoate, trans-methyl pent-2-enoate | |
CC\C=C\C(=O)OC | |
114.14 | |
114 | |
Liquid |
Colorless | |
Authentic | |
C6H10O2 | |
MFCD00137611 | |
MBAHGFJTIVZLFB-SNAWJCMRSA-N | |
methyl (2E)-pent-2-enoate | |
5364718 | |
95% | |
Methyl trans-2-pentenoate, 95% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Methyl trans-2-pentenoate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.