missing translation for 'onlineSavingsMsg'
Learn More

Methyl isothiocyanate, polymer-supported, 1.5-1.9 mmol/g on polystyrene, Thermo Scientific™

Product Code. 11378786
1 g
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-61-6
Molecular Formula C2H3NS
Molecular Weight (g/mol) 73.11
MDL Number MFCD00004818,MFCD03457480
InChI Key LGDSHSYDSCRFAB-UHFFFAOYSA-N
Synonym methyl isothiocyanate, isothiocyanatomethane, methane, isothiocyanato, trapex, methyl mustard oil, methyl mustard, mitc, mencs, methylisothiocyanate, trapexide
PubChem CID 11167
ChEBI CHEBI:78337
IUPAC Name methylimino(sulfanylidene)methane
SMILES CN=C=S

Specifications

Quantity 1 g
Sensitivity Moisture sensitive
Chemical Name or Material Methyl isothiocyanate, polymer-supported, 1.5 to 1.9mmol/g on polystyrene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl isothiocyanate, polymer-supported, 1.5-1.9 mmol/g on polystyrene, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.