Learn More
Methyl 7-bromoheptanoate, 98% Product Code.: 11332239
Methyl 7-bromoheptanoate, 98%
Product Code. 11332239
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11332239
Brand: Thermo Scientific Alfa Aesar H27234.06
Request Bulk or Custom Format
CAS: 54049-24-0 | C8H15BrO2 | 223.11 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54049-24-0
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD02258672
InChI Key BXRLUWIDTDLHQE-UHFFFAOYSA-N
Synonym methyl-7-bromoheptanoate, 7-bromoheptanoic acid methyl ester, heptanoic acid, 7-bromo-, methyl ester, acmc-1aya7, 13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-one
PubChem CID 554086
IUPAC Name methyl 7-bromoheptanoate
SMILES COC(=O)CCCCCCBr
Specifications
Density 1.257
Quantity 5 g
Formula Weight 223.11
Percent Purity 98%
Chemical Name or Material Methyl 7-bromoheptanoate
Product Title
Methyl 7-bromoheptanoate, 98% >