missing translation for 'onlineSavingsMsg'
Learn More
Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™ Product Code.: 10239432

Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10239432
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10239432

Brand: Thermo Scientific Maybridge CC39322EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 118465-49-9
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD07772830
InChI Key YBIAQTPXONLBJG-UHFFFAOYSA-N
Synonym methyl 6h-thieno 2,3-b pyrrole-5-carboxylate, methyl thiopheno 3,2-d pyrrole-5-carboxylate, 5-methoxycarbonyl-6h-thieno 2,3-b pyrroleShow MoreShow Less
PubChem CID 7164601
IUPAC Name methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate
SMILES COC(=O)C1=CC2=C(N1)SC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 181.21
Percent Purity 97%
Chemical Name or Material Methyl 6H-thieno[2, 3-b]pyrrole-5-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.