Learn More
Methyl 4-cyanobenzoate, 99%
Description
It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.
Solubility
Insoluble in water. Soluble in chloroform and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | 65°C to 69°C |
Boiling Point | 142°C to 144°C (12 mmHg) |
Quantity | 5 g |
Beilstein | 1866980 |
Solubility Information | Insoluble in water. Soluble in chloroform and ethyl acetate. |
Formula Weight | 161.16 |
Percent Purity | 99% |
Chemical Name or Material | Methyl 4-cyanobenzoate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
10216302 | Certificate of Analysis | 26/06/2023 | L08365,L08365.06,L08365.14,L08365.22 |
10173786 | Certificate of Analysis | 26/06/2023 | L08365,L08365.06,L08365.14,L08365.22 |
10173786 | Certificate of Analysis | 08/12/2021 | L08365.06,L08365.14,L08365.22 |
10141280 | Certificate of Analysis | 08/12/2021 | L08365.06,L08365.14,L08365.22 |
10165447 | Certificate of Analysis | 06/12/2021 | L08365.06,L08365.14,L08365.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.