missing translation for 'onlineSavingsMsg'
Learn More
Methyl 4-cyanobenzoate, 99%, Thermo Scientific Chemicals Product Code.: 11358115

Methyl 4-cyanobenzoate, 99%, Thermo Scientific Chemicals

Product Code. 11358115
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11358115

Brand: Thermo Scientific Alfa Aesar L08365.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.

Solubility
Insoluble in water. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-35-7
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00001823
InChI Key KKZMIDYKRKGJHG-UHFFFAOYSA-N
Synonym 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5iShow MoreShow Less
PubChem CID 70791
IUPAC Name methyl 4-cyanobenzoate
SMILES COC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 65°C to 69°C
Boiling Point 142°C to 144°C (12 mmHg)
Quantity 25 g
Beilstein 1866980
Solubility Information Insoluble in water. Soluble in chloroform and ethyl acetate.
Formula Weight 161.16
Percent Purity 99%
Chemical Name or Material Methyl 4-cyanobenzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10216302 Certificate TypeCertificate of Analysis Date26/06/2023 Catalog NumberL08365,L08365.06,L08365.14,L08365.22
Lot Number10173786 Certificate TypeCertificate of Analysis Date26/06/2023 Catalog NumberL08365,L08365.06,L08365.14,L08365.22
Lot Number10173786 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08365.06,L08365.14,L08365.22
Lot Number10141280 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08365.06,L08365.14,L08365.22
Lot Number10165447 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL08365.06,L08365.14,L08365.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4-cyanobenzoate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.