Learn More
Methyl 4-acetylbenzoate, 99% Product Code.: 11460167
Methyl 4-acetylbenzoate, 99%
Product Code. 11460167
10 g, 10g
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. 11460167
Brand: Thermo Scientific Alfa Aesar B21626.09
Request Bulk or Custom Format
CAS: 3609-53-8 | C10H10O3 | 178.187 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3609-53-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00216474
InChI Key QNTSFZXGLAHYLC-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl-, methyl ester, unii-3s3d53j4nt, 4-acetylbenzoic acid methyl ester, methyl4-acetylbenzoate, 4-acetyl-benzoic acid methyl ester, methyl 4-acetyl benzoate, pubchem22273, acmc-209ikx, #, ksc497k9b
PubChem CID 137990
IUPAC Name methyl 4-acetylbenzoate
SMILES CC(=O)C1=CC=C(C=C1)C(=O)OC
Specifications
Melting Point 92°C to 95°C
Quantity 10 g
Formula Weight 178.19
Percent Purity 99%
Chemical Name or Material Methyl 4-acetylbenzoate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Product Code
Lot Number10251234 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeB21626.04,B21626.09
Lot Number10175287 Certificate TypeCertificate of Analysis Date02/04/2024 Product CodeB21626.04,B21626.09
Lot Number10175287 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21626.04,B21626.09
Lot NumberI2058A Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21626.04,B21626.09
Product Title
Methyl 4-acetylbenzoate, 99% >
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis