missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylthio)butyrate, 98%, Thermo Scientific™

Product Code. 11425986
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51534-66-8
Molecular Formula C6H12O2S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00013393
InChI Key PBYSEUNXLXTEAN-UHFFFAOYSA-N
PubChem CID 560572
IUPAC Name methyl 2-methylsulfanylbutanoate
SMILES CCC(C(=O)OC)SC

Specifications

Density 1.028
Boiling Point 103°C to 104°C (35 mmHg)
Flash Point 64°C (147°F)
Refractive Index 1.461
Quantity 5 g
Beilstein 1925141
Synonym methyl 2-methylthio butyrate, methyl 2-methylsulfanyl butanoate, butanoic acid,2-methylthio-, methyl ester, methyl 2-methylthio-butyrate, methyl alpha-methylsulfanyl butyrate, acmc-20akj9, 2-methylthio-butanoic acid methyl ester, butanoic acid, 2-methylthio-, methyl ester, +/--butanoic acid, 2-methylthio-, methyl ester, butanoic acid, 2-methylthio-, methyl ester, +/-
Formula Weight 148.23
Percent Purity 98%
Chemical Name or Material Methyl 2-(methylthio)butyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.