missing translation for 'onlineSavingsMsg'
Learn More
Methoxyamine Hydrochloride, MP Biomedicals™ Product Code.: 11477692

Methoxyamine Hydrochloride, MP Biomedicals™

Product Code. 11477692 Shop All MP Biomedicals Products
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11477692

Brand: MP Biomedicals 0215540505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methoxyamine Hydrochloride is used as a reagent for the preparation of O-methyl oximes.

  • Also used to make O-methyl oximes from aldehydes, or ketones.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point 300°F to 304°F (decomposes)
Quantity 5 g
Solubility Information Soluble
Formula Weight 83.52
Physical Form Crystalline Powder
Chemical Name or Material Methoxyamine Hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methoxyamine Hydrochloride, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.