Learn More
SPEX CertiPrep™ Method EN15662 SPEXQuE™ QuEChERS Kit
Meet your QuEChERS needs with these general purpose kits
Brand: SPEX CertiPrep™ EN-PSA-2ML
This item is not returnable.
View return policy
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
- European Methods (EN Method 15662)
Specifications
25 mg PSA, 150 mg MgSO4 | |
2 mL | |
100 Pack |
PSA Dispersive SPE Clean-up Tube | |
EN 15662 |
Product Content Correction
Product Title
SPEX CertiPrep™ Method EN15662 SPEXQuE™ QuEChERS Kit > 100 pk. with 2mL cengtrifuge tube volume