missing translation for 'onlineSavingsMsg'
Learn More

Internal Standard, Acenapthene-d10, SPEX CertiPrep™

Product Code. 16309077
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Acenaphthene-d1015067-26-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 15067-26-2, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.2, 99.8
Quantity 1 mL
CAS Max % 0.2, 99.8
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C12H10
InChI Key CWRYPZZKDGJXCA-WHUVPORUSA-N
SMILES C1CC2=CC=CC3=C2C1=CC=C3
IUPAC Name 1,1,2,2,3,4,5,6,7,8-decadeuterioacenaphthylene
Molecular Weight (g/mol) 164.273
PubChem CID 177559
Methods For US EPA Method 548.1
Analyte or Component Names Acenaphthene-d10
Solvent or Matrix Methanol
Chemical Name or Material Acenapthene-d10
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Internal Standard
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'