missing translation for 'onlineSavingsMsg'
Learn More
Methiothepin mesylate, MedChemExpress Product Code.: 16454158

Methiothepin mesylate, MedChemExpress

Product Code. 16454158
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16454158

Brand: Medchem Express HY107836

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methiothepin mesylate is a potent and non-selective 5-HT2 receptor antagonist, with pKds of 7.10 (5-HT1A), 7.28 (5HT1B), 7.56 (5HT1C), 6.99 (5HT1D), 7.0 (5-HT5A), 7.8 (5-HT5B), 8.74 (5-HT6), and 8.99 (5-HT7), and pKis of 8.50 (5HT2A), 8.68 (5HT2B), and 8.35 (5HT2C).

Methiothepin mesylate is a potent and non-selective 5-HT2 receptor antagonist, with pKds of 7.10 (5-HT1A), 7.28 (5HT1B), 7.56 (5HT1C), 6.99 (5HT1D), 7.0 (5-HT5A), 7.8 (5-HT5B), 8.74 (5-HT6), and 8.99 (5-HT7), and pKis of 8.50 (5HT2A), 8.68 (5HT2B), and 8.35 (5HT2C).

Specifications

Chemical Name or Material Methiothepin mesylate
CAS 74611-28-2
Color White
Purity Grade Notes Research
Molecular Formula C21H28N2O3S3
Quantity 10 mg
Synonym Metitepine mesylate Ro 8-6837 mesylate
Solubility Information DMSO : 62.5 mg/mL (138.08 mM; Need ultrasonic) ∣H2O : 50 mg/mL (110.46 mM; Need ultrasonic)
SMILES CN1CCN(C2CC3=CC=CC=C3SC4=CC=C(SC)C=C24)CC1.CS(=O)(O)=O
Molecular Weight (g/mol) 452.65
Formula Weight 452.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Methiothepin mesylate, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.