missing translation for 'onlineSavingsMsg'
Learn More
Metalaxyl, SPEX CertiPrep™ Product Code.: 16370098
compliance-icons compliance-icons

Metalaxyl, SPEX CertiPrep™

Product Code. 16370098 Shop All SPEX Certiprep Products
1 mL
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy

Product Code. 16370098

Brand: SPEX Certiprep S2369A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Metalaxyl57837-19-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 57837-19-1, 67-64-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Molecular Formula C15H21NO4
InChI Key ZQEIXNIJLIKNTD-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)C)N(C(C)C(=O)OC)C(=O)COC
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
IUPAC Name methyl 2-(N-(2-methoxyacetyl)-2,6-dimethylanilino)propanoate
Molecular Weight (g/mol) 279.336
PubChem CID 42586
ChEBI CHEBI:82790
Solvent or Matrix Acetone
Chemical Name or Material Metalaxyl
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Metalaxyl
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetone, metalaxyl

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metalaxyl, SPEX CertiPrep™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.