Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 2, 3 or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand: Hamilton™ 7748-16
This item is not returnable.
View return policy
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Point 2, 3 or 4 | |
6 Pk. |