missing translation for 'onlineSavingsMsg'
Learn More
Benzo(b)fluoranthene in Acetonitrile 10μg/mL, Fisher Chemical™ Product Code.: 12909613
compliance-icons compliance-icons

Benzo(b)fluoranthene in Acetonitrile 10μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12909613
10 mL
Click to view available options
Quantity:
10 mL
This item is not returnable. View return policy

Product Code. 12909613

Brand: Fisher Chemical P806450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 205-99-2
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C20H12
UN Number 1648
Concentration 10 μg/mL
InChI Key FTOVXSOBNPWTSH-UHFFFAOYSA-N
SMILES C1=CC=C2C3=C4C(=CC=C3)C5=CC=CC=C5C4=CC2=C1
Molecular Weight (g/mol) 252.316
PubChem CID 9153
ChEBI CHEBI:34565
Solvent or Matrix Acetonitrile
Chemical Name or Material Benzo(b)fluoranthene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Benzo(b)fluoranthene 10ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzo(b)fluoranthene in Acetonitrile 10μg/mL, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.