missing translation for 'onlineSavingsMsg'
Learn More
Aroclor 1242 in DCM 100μg/mL, Fisher Chemical™ Product Code.: 12986123

Aroclor 1242 in DCM 100μg/mL, Fisher Chemical™

Product Code. 12986123
1 mL
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy

Product Code. 12986123

Brand: Fisher Chemical F119761

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 53469-21-9
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C12H6Cl4
Solvent or Matrix Dichloromethane
InChI Key ALFHIHDQSYXSGP-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=C(C=C(C=C2)Cl)Cl
IUPAC Name 1,2-dichloro-3-(2,4-dichlorophenyl)benzene
Molecular Weight (g/mol) 291.98
PubChem CID 63103
Chemical Name or Material Aroclor 1242
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aroclor 1242 in DCM 100μg/mL, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.