missing translation for 'onlineSavingsMsg'
Learn More
1,4-Dichlorobenzene D4 in Methanol 2000μg/mL, Fisher Chemical™ Product Code.: 12923004

1,4-Dichlorobenzene D4 in Methanol 2000μg/mL, Fisher Chemical™

Product Code. 12923004 Shop All Fisher Chemical Products
1 mL
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy

Product Code. 12923004

Brand: Fisher Chemical P872380

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 3855-82-1
Physical Form Liquid
Molecular Formula C6H4Cl2
Concentration 2000 ÎĽg/mL
InChI Key OCJBOOLMMGQPQU-RHQRLBAQSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC(=CC=C1Cl)Cl
IUPAC Name 1,4-dichloro-2,3,5,6-tetradeuteriobenzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 151.022
PubChem CID 123202
Quantity 1 mL
Chemical Name or Material 1,4-Dichlorobenzene D4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Dichlorobenzene D4 in Methanol 2000μg/mL, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.