Learn More
Mesityl oxide, 90+%, remainder 4-methyl-4-penten-2-one
Description
Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.
Solubility
Soluble in water(30g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | -59°C |
| Density | 0.856 |
| Boiling Point | 129°C to 130°C |
| Flash Point | 30°C (86°F) |
| Assay Percent Range | ≥90% |
| Odor | honigähnlich |
| Refractive Index | 1.444 |
| Quantity | 1000 mL |
| UN Number | UN1229 |
| Beilstein | 1361550 |
| Show More |
Safety and Handling
- Mesityl oxide, 90+%, remainder 4-methyl-4-penten-2-one
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 4-methyl-3-penten-2-one
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.