Learn More
Scharlab MERKAPTOETANOL F MOL BIOL 50ML
Brand: Scharlab ME00950050
MERKAPTOETANOL F MOL BIOL 50ML
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- 2-Mercaptoethanol, molecular biology grade
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501-Dispose of contents/container to .