Learn More
MP Biomedicals Menhaden Oil Diet, High, Powder
Menhaden oil diet. Contains AIN-76, casein, menhaden oil and mineral and vitamin mix.
Brand:  MP Biomedicals 0296019420
 View more versions of this product
Product Code. 11422941
  • 10544.88 NOK / 20kg
Explore more special offers
Description
Description
Special procedures are required for the proper preparation of menhaden oil diets since these polyunsaturated oils rapidly oxidize upon exposure to air. Under normal conditions, an unacceptable level of oxidation occurs after just three days of exposure to the atmosphere. In order to minimize oxidation, MP Biomedicals packages the diets under nitrogen in sealed bags. It is recommended that these diets be kept frozen and accordingly, the products are shipped on dry ice.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Single-use
20 kg
Refrigerated
0.3048 m
Animal Diet
Powder
Non-sterile
1 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.