Learn More
Cytiva Membra Clear Membrane
Mixed cellulose ester membranes
Brand:  Cytiva 7141-025
1044.81 NOK valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 11355094
  • 1420.00 NOK / Each
In Stock
to see stock.
Explore more special offers
Description
Membranes for asbestos analysis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MCE
100Pack
25 mm
0.8 μm
Membra Clear Membrane