Learn More
Hecht Karl™ Melting Point Determination Tubes
Brand:  Hecht Karl™ 43005010
Product Code. 10033881
  • 126.00 NOK / Pack of 100
Only null left
Estimated Shipment: 24-07-2025
Specifications
Melting Point Determination Tubes
100 in a Glass Vial
90 mm
100 Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hecht Karl™ Melting Point Determination Tubes > Diameter: 1.5mm