Learn More
Honeywell Fluka™ Melamine-13C3 Product Code.: 15941161
Honeywell Fluka™ Melamine-13C3
Product Code. 15941161 Shop All Honeywell Fluka Products
10 mg, 10mg
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 15941161
Brand: Honeywell Fluka™ R135510MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Packaging Glass Bottle
For Use With (Application) TRIAZINES
CAS 1173022-88-2
Product Content Correction
Product Title
Honeywell Fluka™ Melamine-13C3 >