missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51094-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.Specifications
MEK6 | |
Polyclonal | |
Unconjugated | |
MAP2K6 | |
MAP kinase kinase 6, MAP2K6, MAPK/ERK kinase 6, MAPKK 6, MAPKK6, MEK 6, MEK6, MKK6, PRKMK6, SAPKK3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5608 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52564 | |
MAP2K6 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction