missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12775-1-AP-20UL
This item is not returnable.
View return policy
Description
Transcriptional regulator which directly modulates PDPK1 expression, thus promoting survival of pancreatic beta-cells. Also regulates expression of NDFIP1, BNIP3, and CCNG1.Specifications
MEIS3 | |
Polyclonal | |
Unconjugated | |
MEIS3 | |
Homeobox protein Meis3, Meis homeobox 3, Meis1 related protein 2, MEIS3, MRG2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17537, 361514, 56917 | |
-20°C | |
Liquid |
Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97368, Q99687 | |
Meis3 | |
MEIS3 Fusion Protein Ag3471 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction