-
Thermo Scientific ClickSeal Biocontainment Lids
-
Thermo Scientific Outstanding Capacity Small Footprint
-
Thermo Scientific Fiberlite Rotors for Benchtop Applications
-
Thermo Scientific General Purpose Pro Centrifuges Series
-
Thermo Scientific Fiberlite Rotors for Benchtop Applications
-
Thermo Scientific General Purpose Pro Centrifuges Series
-
Thermo Scientific Outstanding Capacity Small Footprint
-
Thermo Scientific ClickSeal Biocontainment Lids
Thermo Scientific™ Megafuge ST4 Plus Centrifuge Series
Faster processing of more samples with a high capacity-to-footprint ratio and simple programming interface, plus the flexibility to run most tubes and bottles on one centrifuge.
Brand: Thermo Scientific™ 75016070
101641.63 NOK valid until 2025-06-30
Use promo code "24069" to get your promotional price.
Description
The ST4/ST4R Plus series centrifuges come standard with an easy-to-use keypad and high contrast LCD display. This high-capacity and versatile unit offers consistent performance and is ideal for routine applications. Quickly and securely swap between 13 unique rotor options with the tool-less Auto-Lock rotor exchange. Available for purchase individually or in convenient application-based packages.
Cell Culture packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 15 mL and 50 mL conical tube adapters.
Blood Tube packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 5/7 mL and 9/10 mL blood tube adapters.
Easy to Use:
- Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
- Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
- Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific™ Auto-Lock™ III Rotor System:
- Secure locking system allows easy push-button installation and exchange of rotors.
- System saves time when switching between applications.
- Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Thermo Scientific™ ClickSeal™ Bucket Sealing System:
- Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
- System has glove-friendly, one-handed open/close capability.
- System is certified by CAMR™ in Porton Down, UK.
Thermo Scientific™ SMARTSpin™ Technology:
- Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
- Technology optimizes safety and improves separations.
Space-Saving Design:
- Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
Energy Efficient:
- System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | |
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | |
3,800 rpm | |
No | |
Direct, Brushless Induction Low Profile | |
Less waste, Energy efficient | |
See individual centrifuge, rotor and accessory data | |
Choices of 9 Accel/10 Decel Rates | |
Cell Culture Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 15mL and 50mL Conical Tube Adapters | |
Up to 9 hr. 59 min. | |
Option to count up or down | |
Centrifuge: 5 Year Limited, Rotor: See Manual | |
208/240 V 50/60 Hz | |
50/60 Hz |
Microprocessor | |
3,374 x g | |
Megafuge | |
Bright LCD Push-Button User Interface | |
Purchase of Thermo Scientific rotor and accessories required for operation | |
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | |
Megafuge ST4 Plus, TX-1000 Cell Culture Package | |
Up to 6 Programs via LCD User Interface buttons | |
Yes | |
ISO 14971, ISO 13485, ISO 9001 | |
Benchtop Centrifuge | |
See individual components of package | |
208/240 V |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Thermo Scientific ClickSeal Biocontainment Lids
-
Thermo Scientific Outstanding Capacity Small Footprint
-
Thermo Scientific Fiberlite Rotors for Benchtop Applications
-
Thermo Scientific General Purpose Pro Centrifuges Series
-
Thermo Scientific Fiberlite Rotors for Benchtop Applications
-
Thermo Scientific General Purpose Pro Centrifuges Series
-
Thermo Scientific Outstanding Capacity Small Footprint
-
Thermo Scientific ClickSeal Biocontainment Lids
Accessories and Services
-
Unity Lab Services™ Thermo Scientific Centrifugation Valida... IQ/OQ English Documents for Refridgerate... Product Code: 11636180
10991.00 NOK / Each -
Unity Lab Services™ Thermo Scientific™ Centrifugation... IQ/OQ Package for Standard Centrifuge Re... Product Code: 15536209
25215.00 NOK / Each -
Unity Lab Services™ Thermo Scientific™ Centrifugation... IQ/OQ/PQ English Documents for Refidgera... Product Code: 15537494
149825.00 NOK / Each -
Unity Lab Services™ Thermo Scientific™ Centrifugation... IQ/OQ/PQ Package for Centrifuge Refriger... Product Code: 15681539
37660.00 NOK / Each -
Unity Lab Services™ Total Care Service Plan for Thermo Scie... Total Care plan for Std Bench Centrifuge... Product Code: 17250434
12775.00 NOK / Each -
Unity Lab Services™ Tech Direct Service Plan for Thermo Sci... Tech Direct plan for Std Bench Centrifug... Product Code: 17270414
6539.00 NOK / Each -
Unity Lab Services™ Total Care Warranty for Thermo Scientif... Total Care Warranty for Std Bench Centri... Product Code: 17280394
6670.00 NOK / Each -
Unity Lab Services™ Extended Warranty Plan for Thermo Scien... Ex. Warranty Service Plan for St. Bencht... Product Code: 17345070
8758.00 NOK / Each -
Unity Lab Services™ Start-up Service (Various Benchtops) Start-up svc (var.s benchtops) Product Code: 17839602
5305.00 NOK / Each -
Unity Lab Services™ Centrifuge Standard Requalification Ser... Requalification Service Centrifuge Stand... Product Code: 30178682
10369.89 NOK / 1 unit
Special Offers
-
Thermo Scientific Sorvall
,
Fisherbrand
,
Thermo Scientific
Make Us an Offer on Centrifuges
Unlock special pricing on top centrifuges. Make us an offer today and upgrade your lab equipment. Limited time only!Promo Code: 21629Expires: 2025-12-31
Days
250Hours
9Minutes
36Seconds
8 - Multiple Manufacturers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Exclusive Promotion on Equipment and Instruments
Take advantage of our limited-time offer on top-quality equipment and instruments to elevate your research and laboratory capabilities.Promo Code: 24069Expires: 2025-06-30
Days
66Hours
9Minutes
36Seconds