Learn More
Buerkle™ Measuring Jug Industrial with Closed Handle Product Code.: 16696553
Estimated Shipment: 04-12-2025
  • ONLINE EXCLUSIVE PRICE
    66.67 NOK
    59.94 NOK / Each
    Save 6.73 NOK
    10% Off
Only null left
Add to basket
Buerkle™ Measuring Jug Industrial with Closed Handle
Product Code. p-7235853 Shop All Bürkle Products
1000 mL, Each
Click to view available options
Capacity (Metric):
1000 mL
5000 mL
Unit Size:
Each
Product Code. 16696553
Brand: Buerkle™ 72041000
  • ONLINE EXCLUSIVE PRICE
    66.67 NOK
    59.94 NOK / Each
    Save 6.73 NOK
    10% Off
Only null left
Estimated Shipment: 04-12-2025
Add to basket
Only null left
Add to basket
Light and handy polypropylene measuring jug is suitable for industrial applications.
The measuring jug has two measuring scales, embossed transparent and with blue lettering. With ergonomic handle, available in closed design for special stability or with open handle for stacking to save space.The measuring jug is made of polypropylene and is therefore highly resistant to acids, bases and chemicals.
  • PP, transparent
  • With embossed graduation printed in blue
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Capacity (Metric) 1000 mL
Height (Metric) 165 mm
Diameter (Metric) 117 mm
Product Content Correction
Product Title
Buerkle™ Measuring Jug Industrial with Closed Handle >