Learn More
Hirschmann™ Measuring flask Class A trapezoidal with glass stopper
Achieve precise standard solutions. Hirschmann™ Measuring flask Class A trapezoidal with glass stopper is standardized to contain or to deliver a precise amount of liquid. Flask has blue graduations.
Brand:  Hirschmann™ 2960275
Product Code. 11728104
  • 697.00 NOK / Pack of 2
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Description
  • Class A
  • Trapezoil
  • Standard and glass stopper
  • Certified conformity
  • Blue graduation
  • Calibrated for IN, DIN EN ISO 1042
Specifications
Specifications
50 mL
140 mm
NS 21/21
0.06%