Learn More
BrandTech™ VACUUBRAND™ ME4C NT Dry Chemistry Diaphragm Pump
Suitable for laboratory tasks such as vacuum filtration, degassing, solid phase extraction and many others
27545.00 NOK
Specifications
Max. Vacuum | 52.5 torr |
---|---|
Horsepower | 0.24 hp |
Includes | Manual |
Motor Speed | 150 to 1800 rpm |
Material | PTFE (Pump Head), Fluoroplastic (Case) |
Product Code | Brand | Electrical Requirements | Plug Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Electrical Requirements | Plug Type | Price | Quantity & Availability | |||||
10725774
![]() |
BrandTech™ VACUUBRAND™
20731200 |
230 V 50/60 Hz | CEE |
27545.00 NOK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
- Engineered to provide 40% higher flowrates at working vacuum level than other competitive pumps with similar free air capacity ratings
- Whisper quite operation at ultimate vacuum
- Less vibration than previous model
- Flowpaths constructed of materials with excellent corrosion resistance (such as PTFE heads and diaphragms
- Eliminates the misting, handling, and disposal of contaminated oil associated with rotary vane pumps
- Service intervals: 10,000 to 15,000 hours
Specifications
52.5 torr | |
Manual | |
PTFE (Pump Head), Fluoroplastic (Case) | |
45 dB(A) | |
11.1 kg | |
2.3/2.6 CFM | |
7.79 in. | |
10 in. | |
9.56 in. | |
Diaphragm Pump | |
2 | |
50°F | |
Oil-Free | |
16 psi |
0.24 hp | |
150 to 1800 rpm | |
ME 4C NT | |
24.47 lb. | |
104°F | |
50/60 Hz | |
198.12 mm | |
254 mm | |
24.28 cm | |
IP40 | |
Chemical-Resistant | |
Filtration and Concentration of Solvents with Low Boiling Point | |
1 |
Product Title
BrandTech™ VACUUBRAND™ ME4C NT Dry Chemistry Diaphragm Pump