missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Max Tube Racks, Short (10 Tubes)

Max Tube Racks, Short (10 Tubes) are designed for use with the Vortex-Genie Max.

1885.00 NOK - 3705.00 NOK

Specifications

Holds Tubes
Tube Size 29 mm dia.
Product Type Tube Rack
Color White
For Use With (Equipment) Vortex-Genie MAX
View More Specs

Products 2
Product Code Brand Packaging Quantity Price Quantity & Availability  
Product Code Brand Packaging Quantity Price Quantity & Availability  
17261762
View Documents
Scientific Industries
SI-MX010S
1 Ea.
1885.00 NOK
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17271762
View Documents
Scientific Industries
SI-MX020S
2/Pk.
3705.00 NOK
Pack of 2
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
Description

Description

  • Accommodates tubes of any material
Specifications

Specifications

Tubes
Tube Rack
Vortex-Genie MAX
29 mm dia.
White
Videos
SDS
Documents

Documents

Product Certifications