Learn More
Honeywell™ Max Single-Use Earplug LS-400 Refill
Click to view available options
Orange, 200 Pairs/Pk
Description
- Soft foam for comfort and performance
- Bell-shaped design for maximum in-ear comfort without backing out of the ear canal, easier to insert
- Smooth, soil-resistant surface prevents dirt or other substances from accumulating on skin or entering ear canal
- Ideal for workers who need to block out loud noise and demand the best protection and comfort available in an earplug
- Europe's most used polyurethane earplug
- Superior noise-blocking performance in a single-use earplug
- Enhanced, contoured shape is easier to insert and stays in for long-wearing comfort
Specifications
Specifications
Color | Orange |
Certifications/Compliance (Europe) | EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: 1219 EC Attestation: 04.03.66 |
Pairs per Package | 200 Pairs/Pk. |
Noise Reduction Rating (NRR) | SNR: 37 dB |
Plug Shape | Bell |
Product Type | Single-use earplug, dispenser refill |
Disposable | Single-use |
Material | Polyurethane |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Honeywell™ Max Single-Use Earplug LS-400 Refill >