Learn More
IKA™ MATRIX ThermoCover
The heatable ThermoCover prevents condensation on the vial lid and wall. At the same time it enables constant reaction conditions.
Brand:  IKA™ 0030001244
Product Code. 16238701
  • 6446.03 NOK / Each
Only null left
Estimated Shipment: 02-07-2025
Description
Suitable for: MATRIX Orbital Delta F1.5 MATRIX Orbital Delta FP MATRIX Orbital Delta F0.5 MATRIX Orbital Delta F2.0 MATRIX Orbital Delta plus MATRIX Delta plus
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Heatable ThermoCover
190 x 180 x 120 mm
Preventing the formation of condensate on vessel walls/vessel lids
MATRIX Orbital Delta F1.5, MATRIX Orbital Delta FP, MATRIX Orbital Delta F0.5, MATRIX Orbital Delta F2.0, MATRIX Orbital Delta Plus, MATRIX Delta Plus
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IKA™ MATRIX ThermoCover > ThermoCover