Learn More
Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
BIOPHARMAGREENER_CHOICE
Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.
195.48 NOK - 4555.00 NOK
Specifications
Fit for Purpose Sterile and PCR Ready
View More Specs
Products 21
Product Code Brand Color Product Type Price Quantity & Availability  
Product Code Brand Color Product Type Price Quantity & Availability  
11902261
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470BLK
Black Screw Cap
2350.00 NOK
Pack of 500
Only null left
Estimated Shipment: 01-07-2025
Add to basket
11932261
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470ORG
Orange Screw Cap
2345.00 NOK
Pack of 500
Only null left
Estimated Shipment: 01-07-2025
Add to basket
11942261
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470PIN
Pink Screw Cap
2350.00 NOK
Pack of 500
Only null left
Estimated Shipment: 01-07-2025
Add to basket
11865482
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477GRY
Gray Screw Cap Tray
4555.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
11875482
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477PIN
Pink Screw Cap Tray
4555.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
10736554
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470
Colorless Screw Cap
2310.00 NOK
Pack of 500
Only null left
In Stock
Add to basket
11632000
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470RED
Red Screw Cap
2350.00 NOK
Pack of 500
Only null left
In Stock
Add to basket
10780140
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470YEL
Yellow Screw Cap
2350.00 NOK
Pack of 500
Only null left
Estimated Shipment: 10-06-2025
Add to basket
11681970
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470GRE
Green Screw Cap
2350.00 NOK
Pack of 500
Only null left
In Stock
Add to basket
10401908
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470WHI
White Screw Cap
2350.00 NOK
Pack of 500
Only null left
In Stock
Add to basket
11651990
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470PUR
Purple Screw Cap
2310.00 NOK
Pack of 500
Only null left
Estimated Shipment: 18-09-2025
Add to basket
10040273
View Documents
Thermo Scientific™
4471
Red Screw Cap Insert This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11577429
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4906
Yellow Screw Cap Tray, Empty
1055.00 NOK
Pack of 5
Only null left
In Stock
Add to basket
12865601
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477
Colorless Screw Cap Tray
4475.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
11642000
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477RED
Red Screw Cap Tray
4555.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
11612010
BIOPHARMAGREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477YEL
Yellow Screw Cap Tray
4475.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
10290740
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477BLU
Blue Screw Cap Tray
4475.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
11317201
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477GRE
Green Screw Cap Tray
4555.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
11895482
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477WHI
White Screw Cap Tray
4555.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
11377201
BIOPHARMAGREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4477PUR
Purple Screw Cap Tray
4555.00 NOK
Pack of 480
Only null left
In Stock
Add to basket
11601970
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4470BLU
Blue Screw Cap
2350.00 NOK
Pack of 500
Only null left
Estimated Shipment: 18-09-2025
Add to basket
Description
Description
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Highest Quality
  • All caps and inserts are supplied free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Superior Cap Design
  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -176°C
Cap Options Simplify Sample Identification
  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors
Specifications
Specifications
Sterile and PCR Ready
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts