Learn More
3M™ Mat Edging Roll
A vinyl edging designed to match 3M™ Matting. Meets the needs of providing a smooth transition from the flooring to the matting.
Brand: 3M™ 7000028465
This item is not returnable.
View return policy
Description
- For use on vinyl matting to provide an edge that tapers to the floor
Specifications
12 x 0.038 m | |
High Profile | |
12 m | |
0.038 m |
Creating an edge that tapers to the floor | |
Black | |
Vinyl |