Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Head Mounting Hardware
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
Brand: Masterflex™ 77200-02
818.17 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
- For use with L/S Easy-Load II Pump Heads
- See individual drive specifications for maximum number of heads to be mounted
- Mounting hardware is stainless steel
Specifications
Mounting two L/S Easy-Load II pump heads | |
L/S Easy-Load II pump heads | |
Mounting Hardware | |
Stainless Steel |