Learn More
MAP2K4 Mouse anti-Human, Unconjugated, Clone: 2E4C1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67333-1-IG-150UL
 View more versions of this product
Product Code. 16839905
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
The MAP2K4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAP2K4. This antibody recognizes human antigen. The MAP2K4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MAP2K4
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry
2E4C1
Unconjugated
150 μL
6416
IgG2b