Learn More
MAP2K4 Mouse anti-Human, Unconjugated, Clone: 2E4C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67333-1-IG-150UL
This item is not returnable.
View return policy
Description
The MAP2K4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAP2K4. This antibody recognizes human antigen. The MAP2K4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
MAP2K4 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry | |
2E4C1 | |
Unconjugated | |
150 μL | |
6416 | |
IgG2b |