Learn More
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable
Manual decrimping tools allow easy removal of aluminum seals without breakage.
2370.00 NOK - 11265.00 NOK
Specifications
Operation | Manual |
---|---|
Type | Manual Decapper |
For Use With (Equipment) | Aluminum Cap |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11758276
|
Fisherbrand™
08 07 0001 |
8 mm |
3040.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11768276
|
Fisherbrand™
11 07 0002 |
11 mm |
2370.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11787597
|
Fisherbrand™
13 07 0003 |
13 mm |
3040.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11500535
|
Fisherbrand™
20 07 0004 |
20 mm |
2975.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11849440
|
Fisherbrand™
28 07 0092 |
28 mm |
11265.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10621210
|
Fisherbrand™
32 07 0078 |
32 mm |
11265.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Decapper
- Manual
- Steel
Specifications
Manual | |
Manual Decapper | |
Aluminum Cap |
Product Title
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable