Learn More
Thermo Scientific Chemicals Maltitol, 95%
CAS: 585-88-6 | C12H24O11 | 344.32 g/mol
422.00 NOK - 4150.00 NOK
Chemical Identifiers
CAS | 585-88-6 |
---|---|
Molecular Formula | C12H24O11 |
Molecular Weight (g/mol) | 344.32 |
MDL Number | MFCD00006600 |
InChI Key | VQHSOMBJVWLPSR-WUJBLJFYSA-N |
Synonym | maltitol, d-maltitol, maltisorb, malbit, amalti syrup, malti mr, amalty mr 100, amalty, 4-o-alpha-d-glucopyranosyl-d-glucitol, unii-d65dg142wk |
PubChem CID | 493591 |
ChEBI | CHEBI:68428 |
IUPAC Name | (2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol |
SMILES | C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10773451
|
Thermo Scientific Chemicals
295800250 |
25 g | Glass bottle |
422.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10780571
|
Thermo Scientific Chemicals
295802500 |
250 g | Glass bottle |
1885.00 NOK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10570994
|
Thermo Scientific Chemicals
295800010 |
1 kg | Glass bottle |
4150.00 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
585-88-6 | |
344.32 | |
VQHSOMBJVWLPSR-WUJBLJFYSA-N | |
493591 | |
(2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol |
C12H24O11 | |
MFCD00006600 | |
maltitol, d-maltitol, maltisorb, malbit, amalti syrup, malti mr, amalty mr 100, amalty, 4-o-alpha-d-glucopyranosyl-d-glucitol, unii-d65dg142wk | |
CHEBI:68428 | |
C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O |
Specifications
585-88-6 | |
White | |
94% min. (ELSD) (HPLC) | |
MFCD00006600 | |
+ 107.00 (20.00°C c=5,water) | |
maltitol, d-maltitol, maltisorb, malbit, amalti syrup, malti mr, amalty mr 100, amalty, 4-o-alpha-d-glucopyranosyl-d-glucitol, unii-d65dg142wk | |
VQHSOMBJVWLPSR-WUJBLJFYSA-N | |
(2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol | |
493591 | |
344.32 | |
Crystalline Powder |
147°C to 153°C | |
Authentic | |
C12H24O11 | |
17, V,7, 145 | |
+ 107 | |
(10% in water) Clear colorless | |
C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O | |
344.32 | |
CHEBI:68428 | |
95% | |
Maltitol, 95% |
Safety and Handling
missing translation for 'einecsNumber' : 209-567-0
missing translation for 'rtecsNumber' : LZ4394000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.