Learn More
Thermo Scientific™ Malt Extract Agar (Dehydrated)
Detect, isolate and enumerate yeasts and molds with typical morphology and pigmentation using medium.
2195.00 NOK
Specifications
Form Powder
Product Type Agar
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10321063
View Documents Promotion Details
Thermo Scientific™
CM0059B
500 g
2195.00 NOK
500g
Description
Description
Detect, isolate and enumerate yeasts and molds with Thermo Scientific™ Oxoid™ Malt Extract Agar (Dehydrated). Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation.
Malt Extract Agar is similar to the formula of Galloway and Burgess1 used for the detection, isolation, and enumeration of yeasts and molds. For mycological counts, it may be desirable to prepare the more acid medium in order to suppress bacterial growth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
Powder
Agar
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Product Certifications
For Laboratory Use Only
Product Content Correction
Product Title
Thermo Scientific™ Malt Extract Agar (Dehydrated)