Learn More
Eppendorf™ Mains/Power Cord, 230 V, 50 Hz (EU)
Power cord: 230 V, 50 Hz (EU)
Brand:  Eppendorf™ 0013563934
230.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 30040078
  • 230.92 NOK / Each
Only null left
In Stock
Description
  • 230 V
  • 50 Hz
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Power Cord
230 V/50 Hz (EU)
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Mains/Power Cord, 230 V, 50 Hz (EU) > 230 V/50 Hz (EU)