Learn More
Grant Instruments™ Mini Magnetic Stirrer
MMS-3000 Compact mini magnetic stirrer for routine laboratory procedures.
Brand: Grant Instruments™ MMS-3000
3054.63 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
With its quiet operation and small size, its fits neatly into most laboratory workspaces.- Adjustable speed control: 0 to 3000 rpm
- Handles volumes up to 20 liters
- Supplied with a stirrer rod
- Quiet operation, small size - fits neatly into the workspace
- Rugged stainless steel plate provides a working surface of 110 x 110mm
- Easy-to-use graduated dial
- Two-year warranty
Specifications
0 to 3000 rpm | |
120/230 V | |
230 mm | |
1.5 kg |
20 L | |
75 mm | |
185 mm |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
25Hours
18Minutes
29Seconds
34
Product Content Correction
Product Title
Grant Instruments™ Mini Magnetic Stirrer > Speed: 0 to 3000rpm